Läs mer
Hydrocortisone 21-Hemisuccinate, TRC
Produktkod. 30135276
Change view
Kvantitet:
1 g
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135276 2.5 g
30005468 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135276 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH7165632.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2203-97-6
Molekylformel C25 H34 O8
Molekylvikt (g/mol) 462.53
Synonym Hydrocortisone hydrogen succinate, 11β,17-Dihydroxy-3,20-dioxopregn-4-en-21-yl hydrogen butanedioate, Pregn-4-ene-3,20-dione, 21-(3-carboxy-1-oxopropoxy)-11,17-dihydroxy-, (11β)-, Cortisol, 21-(hydrogen succinate) (8CI), Cortisol, hydrogen succinate (7CI), (11β)-21-(3-Carboxy-1-oxopropoxy)-11,17-dihydroxypregn-4-ene-3,20-dione, Cortisol 21-hemisuccinate, Cortisol hemisuccinate, Cortisol succinate, Hydrocortisone 21-(hydrogen succinate), Hydrocortisone 21-O-β-carbonylpropionic acid, Hydrocortisone 21-hemisuccinate, Hydrocortisone 21-succinate, Hydrocortisone hemisuccinate, Hydrocortisone succinate, NSC 7576, Saxizon
IUPAC-namn 4-[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoic acid
LEDER C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COC(=O)CCC(=O)O)[C@@]4(C)C[C@H](O)[C@H]23
Specifikationer
Kemiskt namn eller material Hydrocortisone Hydrogen Succinate
Kvantitet 2.5 g
InChI formel InChI=1S/C25H34O8/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30/h11,16-18,22,27,32H,3-10,12-13H2,1-2H3,(H,29,30)/t16-,17-,18-,22+,23-,24-,25-/m0/s1
Formel vikt 462.2254