Läs mer
Hydrocortisone 21-Propionate, TRC
Produktkod. 30138399
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138399 25 mg
30009145 50 mg
30006088 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138399 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH71477525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6677-98-1
Molekylformel C24 H34 O6
Molekylvikt (g/mol) 418.52
Synonym Pregn-4-ene-3,20-dione, 11,17-dihydroxy-21-(1-oxopropoxy)-, (11β)-, Cortisol, 21-propionate (6CI,7CI,8CI), Hydrocortisone 21-propionate, Hydrocortisone propionate
IUPAC-namn [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
LEDER CCC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material Hydrocortisone 21-Propionate
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C24H34O6/c1-4-20(28)30-13-19(27)24(29)10-8-17-16-6-5-14-11-15(25)7-9-22(14,2)21(16)18(26)12-23(17,24)3/h11,16-18,21,26,29H,4-10,12-13H2,1-3H3/t16-,17-,18-,21+,22-,23-,24-/m0/s1
Formel vikt 418.2355
Procent renhet >95%