Läs mer
Hydrocortisone Hemisuccinate Hydrate, TRC
Produktkod. 30137349
Change view
Kvantitet:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137349 1 g
30133624 100 mg
30007800 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137349 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH7147651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83784-20-7
Molekylformel C25 H34 O8 . H2 O
Molekylvikt (g/mol) 480.55
Synonym Pregn-4-ene-3,20-dione, 21-(3-carboxy-1-oxopropoxy)-11,17-dihydroxy-, hydrate (1:1), (11β)-, Pregn-4-ene-3,20-dione, 21-(3-carboxy-1-oxopropoxy)-11,17-dihydroxy-, monohydrate, (11β)- (9CI), Hydrocortisone hydrogen succinate monohydrate, Hydrocortisone hemisuccinate monohydrate
IUPAC-namn 4-[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoic acid;hydrate
LEDER O.C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COC(=O)CCC(=O)O)[C@@]4(C)C[C@H](O)[C@H]23
Specifikationer
Kemiskt namn eller material Hydrocortisone hemisuccinate hydrate
Kvantitet 1 g
InChI formel InChI=1S/C25H34O8.H2O/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30;/h11,16-18,22,27,32H,3-10,12-13H2,1-2H3,(H,29,30);1H2/t16-,17-,18-,22+,23-,24-,25-;/m0./s1
Formel vikt 480.2359