Läs mer
Hydrocortisone Sodium Phosphate, TRC
Produktkod. 30136498
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136498 10 g
30010076 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136498 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH71469010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6000-74-4
Molekylformel C21 H29 O8 P . 2 Na
Molekylvikt (g/mol) 486.4
Synonym Pregn-4-ene-3,20-dione, 11,17-dihydroxy-21-(phosphonooxy)-, sodium salt (1:2), (11β)-, Cortisol 21-phosphate disodium salt (6CI), Cortisol, 21-(dihydrogen phosphate), disodium salt (7CI,8CI), Pregn-4-ene-3,20-dione, 11,17-dihydroxy-21-(phosphonooxy)-, disodium salt, (11β)- (9CI), Cleiton, Cortisol sodium phosphate, Disodium hydrocortisone 21-phosphate, Efcortesol, Hydrocortisone 21-disodium phosphate, Hydrocortisone 21-disodium salt, Hydrocortisone 21-phosphate disodium, Hydrocortisone 21-phosphate disodium salt, Hydrocortisone 21-sodium phosphate, Hydrocortisone disodium phosphate, Hydrocortisone sodium phosphate, Hydrocortone phosphate, Physiocortison, Sodium cortisol 21-phosphate, Sodium hydrocortisone 21-phosphate
IUPAC-namn disodium;[2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate
LEDER [Na+].[Na+].C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COP(=O)([O-])[O-])[C@@]4(C)C[C@H](O)[C@H]23
Specifikationer
Kemiskt namn eller material Hydrocortisone Sodium Phosphate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C21H31O8P.2Na/c1-19-7-5-13(22)9-12(19)3-4-14-15-6-8-21(25,17(24)11-29-30(26,27)28)20(15,2)10-16(23)18(14)19;;/h9,14-16,18,23,25H,3-8,10-11H2,1-2H3,(H2,26,27,28);;/q;2*+1/p-2/t14-,15-,16-,18+,19-,20-,21-;;/m0../s1
Formel vikt 486.1395
Procent renhet >95%