Läs mer
Hydrogenated Soya Phosphatidylcholine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Hydrogenated Soya Phosphatidylcholine |
| CAS | 97281-48-6 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C44H88NO8P |
| Kvantitet | 1 g |
| Synonym | Coatsome NC 21E, HSPC, Hydrogenated Soy Phosphatidylcholines, NC 21E, Phospholipon 90N |
| InChI formel | InChI=1S/C44H88NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h42H,6-41H2,1-5H3 |
| LEDER | CCCCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCCCC |
| Molekylvikt (g/mol) | 790.1 |
| Procent renhet | >95 |