Läs mer
Hydroquinidine, MedChemExpress
Beskrivning
Hydroquinidine (Dihydroquinidine) is a derivative of Quinidine (an antiarrhythmic agent). Hydroquinidine prolongs the QT interval and has antiarrhythmic efficacy.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Hydroquinidine |
| CAS | 1435-55-8 |
| Färg | White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C20H26N2O2 |
| Kvantitet | 100 mg |
| Synonym | Dihydroquinidine (+)-Hydroquinidine Hydroconquinine |
| Löslighetsinformation | DMSO : 20.83 mg/mL (63.81 mM; Need ultrasonic) |
| LEDER | O[C@@H](C1=CC=NC2=C1C=C(OC)C=C2)[C@]3([H])[N@](C[C@@H]4CC)CC[C@H]4C3 |
| Molekylvikt (g/mol) | 326.43 |
| Visa mer |