Läs mer
Hydroxy Acetildenafil, TRC
Produktkod. 30009345
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009345 50 mg
17902995 10 mg
17931265 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH73994050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 147676-56-0
Molekylformel C25 H34 N6 O4
Molekylvikt (g/mol) 482.58
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[2-[4-(2-hydroxyethyl)-1-piperazinyl]acetyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[[4-(2-hydroxyethyl)-1-piperazinyl]acetyl]phenyl]-1,4-dihydro-1-methyl-3-propyl- (9CI), Hydroxyhongdenafil
IUPAC-namn 5-[2-ethoxy-5-[2-[4-(2-hydroxyethyl)piperazin-1-yl]acetyl]phenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
LEDER CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)C(=O)CN4CCN(CCO)CC4
Specifikationer
Kemiskt namn eller material Hydroxyacetildenafil
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C25H34N6O4/c1-4-6-19-22-23(29(3)28-19)25(34)27-24(26-22)18-15-17(7-8-21(18)35-5-2)20(33)16-31-11-9-30(10-12-31)13-14-32/h7-8,15,32H,4-6,9-14,16H2,1-3H3,(H,26,27,34)
Formel vikt 482.2642
Procent renhet >95%