Läs mer
Hydroxy Adapalene, TRC
Produktkod. 17914045
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17914045 25 mg
17963175 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17914045 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH75004525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1346599-76-5
Molekylformel C28H28O4
Molekylvikt (g/mol) 428.52
Synonym 6-[3-(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)-4-methoxyphenyl]-2-naphthalenecarboxylic Acid
LEDER COC1=C(C=C(C=C1)C2=CC3=C(C=C2)C=C(C=C3)C(=O)O)C45CC6CC(C4)CC(C6)(C5)O
Specifikationer
Kemiskt namn eller material Hydroxy Adapalene
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C28H28O4/c1-32-25-7-6-22(20-2-3-21-10-23(26(29)30)5-4-19(21)9-20)11-24(25)27-12-17-8-18(13-27)15-28(31,14-17)16-27/h2-7,9-11,17-18,31H,8,12-16H2,1H3,(H,29,30)
Procent renhet >95%