Läs mer
Hydroxy Meprobamate, TRC
Produktkod. 30135413
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135413 100 mg
30008349 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135413 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH945100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3567-43-9
Molekylformel C9H18N2O5
Molekylvikt (g/mol) 234.25
Synonym 2-[[(Aminocarbonyl)oxy]methyl]-2-methyl-1,4-pentanediol 1-Carbamate, Carbamic Acid 2-(2-Hydroxypropyl)-2-methyltrimethylene Ester;
IUPAC-namn [2-(carbamoyloxymethyl)-4-hydroxy-2-methylpentyl] carbamate
LEDER CC(CC(C)(COC(=O)N)COC(=O)N)O
Specifikationer
Kemiskt namn eller material Hydroxy Meprobamate
InChI formel InChI=1S/C9H18N2O5/c1-6(12)3-9(2,4-15-7(10)13)5-16-8(11)14/h6,12H,3-5H2,1-2H3,(H2,10,13)(H2,11,14)
Kvantitet 100 mg
Formel vikt 234.1216