Läs mer
Hydroxy Terbinafine, TRC
Produktkod. 30005823
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005823 10 mg
30003428 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005823 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95616010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 162227-13-6
Molekylformel C21 H25 N O
Molekylvikt (g/mol) 307.43
Synonym 5-Hepten-3-yn-1-ol, 2,2-dimethyl-7-[methyl(1-naphthalenylmethyl)amino]-, (5E)-, 5-Hepten-3-yn-1-ol, 2,2-dimethyl-7-[methyl(1-naphthalenylmethyl)amino]-, (E)-, (5E)-2,2-Dimethyl-7-[methyl(1-naphthalenylmethyl)amino]-5-hepten-3-yn-1-ol, Hydroxyterbinafine
IUPAC-namn (E)-2,2-dimethyl-7-[methyl(naphthalen-1-ylmethyl)amino]hept-5-en-3-yn-1-ol
LEDER CN(C\C=C\C#CC(C)(C)CO)Cc1cccc2ccccc12
Specifikationer
Kemiskt namn eller material Hydroxy Terbinafine
Kvantitet 10 mg
InChI formel InChI=1S/C21H25NO/c1-21(2,17-23)14-7-4-8-15-22(3)16-19-12-9-11-18-10-5-6-13-20(18)19/h4-6,8-13,23H,15-17H2,1-3H3/b8-4+
Formel vikt 307.1936