Läs mer
Hydroxy Urea-13C,15N2, TRC
Produktkod. 18051520
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18051520 10 mg
18072520 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18051520 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH99100210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246814-92-5
Molekylformel 13CH415N2O2
Molekylvikt (g/mol) 79.03
Synonym N-Hydroxyurea-13C,15N2, Biosupressin-13C,15N2, Carbamohydroxamic Acid-13C,15N2, Carbamoyl Oxime-13C,15N2, Cytodrox-13C,15N2, Droxia-13C,15N2, Hydrea-13C,15N2, Hydreia-13C,15N2, Hydroxycarbamide-13C,15N2, Hydroxycarbamine-13C,15N2, N-(Aminocarbonyl)hydroxylamine-13C,15N2, Hydroxylurea-13C,15N2, Hydura-13C,15N2, Litaler-13C,15N2, N-Carbamoylhydroxylamine-13C,15N2, NSC 32065-13C,15N2, Oxyurea-13C,15N2, SK 22591-13C,15N2, SQ 1089-13C,15N2;
IUPAC-namn 1-hydroxyurea-13C-15N2
LEDER [15NH2][13C]([15NH]O)=O
Specifikationer
Kemiskt namn eller material Hydroxy Urea-13C,15N2
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/CH4N2O2/c2-1(4)3-5/h5H,(H3,2,3,4)/i1+1,2+1,3+1
Formel vikt 79.02
Procent renhet >95%