Läs mer
Hydroxybupropion, Lipomed
Produktkod. 30461396
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30461396 100 mg
1 options
Produktkod. 30461396 Leverantör Lipomed Leverantörsnummer LPMHBP1895FB100
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Hydroxybupropion
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (±)-Hydroxybupropion
CAS 92264-81-8
Analyt- eller komponentnamn (±)-Hydroxybupropion
Frakt skick Room Temperature
Molekylformel C13 H18 Cl N O2
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
LEDER CC(NC(C)(C)CO)C(=O)c1cccc(Cl)c1
IUPAC-namn 1-(3-chlorophenyl)-2-[(1-hydroxy-2-methylpropan-2-yl)amino]propan-1-one
Industry Type Pharmaceutical
Molekylvikt (g/mol) 255.74
Kvantitet 100 mg
Formel vikt 255.103 g/mol