Läs mer
Hydroxychloroquine 3-Carbonitrile, TRC
Produktkod. 30180301
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180301 10 mg
30179992 50 mg
30180243 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180301 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH78393510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2054858-84-1
Molekylformel C19H25ClN4O
Molekylvikt (g/mol) 360.88
Synonym 3-Cyano-Hydroxychloroquine, 7-Chloro-4-[[4-[ethyl(2-hydroxyethyl)amino]-1-methylbutyl]amino]-3-quinolinecarbonitrile
IUPAC-namn 7-chloro-4-[5-[ethyl(2-hydroxyethyl)amino]pentan-2-ylamino]quinoline-3-carbonitrile
LEDER CCN(CCO)CCCC(C)Nc1c(cnc2cc(Cl)ccc12)C#N
Specifikationer
Kemiskt namn eller material Hydroxychloroquine 3-Carbonitrile
Kvantitet 10 mg
InChI formel InChI=1S/C19H25ClN4O/c1-3-24(9-10-25)8-4-5-14(2)23-19-15(12-21)13-22-18-11-16(20)6-7-17(18)19/h6-7,11,13-14,25H,3-5,8-10H2,1-2H3,(H,22,23)
Formel vikt 360.1717