missing translation for 'onlineSavingsMsg'
Läs mer

Hydroxyethylcellulose, Sigma-Aldrich™

Produktkod. 15841651
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 g
500 g
Förpackningsstorlek:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15841651 500 g 500g
15831651 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15841651 Leverantör Sigma Aldrich Leverantörsnummer 8220680500
 Begränsad artikel
Beräknad leverans: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-62-0

Specifications

MDL-nummer MFCD00072770
Kvantitet 500 g
Synonym Hydroxyethylcellulose, Hydroxyethylcellulose
Kvalitet Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.