Läs mer
Hydroxyhomo Sildenafil, TRC
Produktkod. 30010282
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010282 25 mg
17934485 10 mg
17964165 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010282 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94284025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139755-85-4
Molekylformel C23 H32 N6 O5 S
Molekylvikt (g/mol) 504.6
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[[4-(2-hydroxyethyl)-1-piperazinyl]sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, 1-Piperazineethanol, 4-[[3-(4,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]- (9CI), Hydroxyhomosildenafil, Lodenafil
IUPAC-namn 5-[2-ethoxy-5-[4-(2-hydroxyethyl)piperazin-1-yl]sulfonylphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
LEDER CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(CCO)CC4
Specifikationer
Kemiskt namn eller material Hydroxyhomosildenafil
Kvantitet 25 mg
InChI formel InChI=1S/C23H32N6O5S/c1-4-6-18-20-21(27(3)26-18)23(31)25-22(24-20)17-15-16(7-8-19(17)34-5-2)35(32,33)29-11-9-28(10-12-29)13-14-30/h7-8,15,30H,4-6,9-14H2,1-3H3,(H,24,25,31)
Formel vikt 504.2155