missing translation for 'onlineSavingsMsg'
Läs mer

Hydroxylamine, 50% aq. soln., Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar B22202.AE

Kvantitet 100mL

 Se fler versioner av denna produkt

Produktkod. 15453237

Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »

Denna artikel kan inte returneras. Se returpolicy

Beskrivning

Beskrivning

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Hydroxylamine is a reactant used for the preparation of hydroxyaminoguanidines which are anti-cancer agents, carboxamide derivatives of ofloxacin and human immunodeficiency virus (HIV) integrase inhibitors. It reacts with chlorosulfonic acid to give hydroxylamine-O-sulfonic acid, a useful reagent in the synthesis of caprolactam. It is commonly used as a reducing agent in myriad of organic and inorganic reactions. It acts as an antioxidant for fatty acids. It is widely used to prepare oximes, an important functional group.

Solubility
Miscible with water, liquid ammonia and methanol. Slightly miscible with ether, benzene, carbon disulfide and chloroform.

Notes
It can violently decompose at elevated temperatures. May be shock-sensitive if dry. Do not heat over 500C. Incompatible with bases, oxidizing agents, nitrates, nitrites, heavy metals, rubber, metals, epoxides and polyester.
TRUSTED_SUSTAINABILITY
Specifikationer

Kemiska identifierare

7803-49-8
33.03
AVXURJPOCDRRFD-UHFFFAOYSA-N
787
hydroxylamine
H3NO
MFCD00044522
oxammonium, nitroxide, hydroxyamine, nh2oh, oxyammonia, nitrinous acid, unii-2fp81o2l9z, hsdb 579, dihydridohydroxidonitrogen, arninoalcohol
CHEBI:15429
NO

Specifikationer

Hydroxylamine
7803-49-8
0.5
NH2OH
UN3266
None
oxammonium, nitroxide, hydroxyamine, nh2oh, oxyammonia, nitrinous acid, unii-2fp81o2l9z, hsdb 579, dihydridohydroxidonitrogen, arninoalcohol
AVXURJPOCDRRFD-UHFFFAOYSA-N
hydroxylamine
787
33.03
1.120
aq. soln.
100mL
H3NO
MFCD00044522
1098214
14,4828
Miscible with water,liquid ammonia and methanol. Slightly miscible with ether,benzene,carbon disulfide and chloroform.
NO
33.03
CHEBI:15429
1.394
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Certifikat
Kampanjer

Kampanjer

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.