Läs mer
Hydroxylysylpyridinoline, TRC
Produktkod. 30007323
Change view
Kvantitet:
0.5 mg
5 mg
Förpackningsstorlek:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30007323 0.5 mg 0.5mg
30006773 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007323 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9540360.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 63800-01-1
Molekylformel C18H28N4O8
Molekylvikt (g/mol) 428.44
Synonym 2-Amino-6-[4-[(2S)-2-amino-2-carboxyethyl]-3-[(3S)-3-amino-3-carboxypropyl]-5-hydroxypyridinio]-2,3,4,6-tetradeoxy-L-erythro-Hexonic Acid Inner Salt, Pyridinoline
IUPAC-namn 2-amino-6-[4-(2-amino-2-carboxyethyl)-3-(3-amino-3-carboxypropyl)-5-hydroxypyridin-1-ium-1-yl]-5-hydroxyhexanoate
LEDER C1=C(C(=C(C=[N+]1CC(CCC(C(=O)[O-])N)O)O)CC(C(=O)O)N)CCC(C(=O)O)N
Specifikationer
Kemiskt namn eller material Hydroxylysylpyridinoline
Anteckningar om renhetsgrad HPLC
Kvantitet 0.5 mg
InChI formel InChI=1S/C18H28N4O8/c19-12(16(25)26)3-1-9-6-22(7-10(23)2-4-13(20)17(27)28)8-15(24)11(9)5-14(21)18(29)30/h6,8,10,12-14,23H,1-5,7,19-21H2,(H3-,24,25,26,27,28,29,30)
Formel vikt 428.191
Procent renhet >95%