Läs mer
Hydroxymethyl Tolperisone Hydrochloride, TRC
Produktkod. 30407477
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30407477 10 mg
1 options
Produktkod. 30407477 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH82552510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hydroxymethyl Tolperisone Hydrochloride
CAS 352233-14-8
Molekylformel C16 H23 N O2 . Cl H
Kvantitet 10 mg
Synonym 1-[4-(Hydroxymethyl)phenyl]-2-methyl-3-(1-piperidinyl)-1-propanone Hydrochloride
InChI formel InChI=1S/C16H23NO2.ClH/c1-13(11-17-9-3-2-4-10-17)16(19)15-7-5-14(12-18)6-8-15;/h5-8,13,18H,2-4,9-12H2,1H3;1H
LEDER Cl.CC(CN1CCCCC1)C(=O)c2ccc(CO)cc2
IUPAC-namn 1-[4-(hydroxymethyl)phenyl]-2-methyl-3-(1-piperidyl)propan-1-one;hydrochloride
Molekylvikt (g/mol) 297.82
Formel vikt 297.1496