Läs mer
Hydroxyphosphonoacetic Acid - 50% in water, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Hydroxyphosphonoacetic Acid |
| CAS | 23783-26-8 |
| Molekylformel | C2 H5 O6 P |
| Kvantitet | 1 g |
| Synonym | 2-Hydroxy-2-phosphonoacetic Acid, 2-Hydroxyphosphonoacetic Acid, 2-Hydroxyphosphonocarboxylic Acid, Belcor 575, HPPA (scale inhibitor), Hydroxyphosphonoacetic Acid, Phosphonoglycolic Acid, Phosphonohydroxyacetic Acid, α-Hydroxyphosphonoacetic Acid |
| InChI formel | InChI=1S/C2H5O6P/c3-1(4)2(5)9(6,7)8/h2,5H,(H,3,4)(H2,6,7,8) |
| LEDER | OC(C(=O)O)P(=O)(O)O |
| IUPAC-namn | 2-hydroxy-2-phosphonoacetic acid |
| Molekylvikt (g/mol) | 156.031 |
| Formel vikt | 155.982 |
| Visa mer |