Läs mer
Hydroxypropyl Chitosan, TRC
Produktkod. 30132598
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132598 1 g
17957634 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132598 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9527401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84069-44-3
Molekylformel C36H72N2O17[C15H29NO7]n
Molekylvikt (g/mol) 1140.353
Synonym Chitofilmer, 2-Hydroxypropyl Ether Chitosan
IUPAC-namn 1-[[(2R,5S)-5-[(2S,5S)-5-[(2S,5S)-4,5-bis(2-hydroxypropoxy)-6-(2-hydroxypropoxymethyl)-3-(2-hydroxypropylamino)oxan-2-yl]oxy-4-(2-hydroxypropoxy)-6-(2-hydroxypropoxymethyl)-3-(2-hydroxypropylamino)oxan-2-yl]oxy-2,4-bis(2-hydroxypropoxy)-6-(2-hydroxypropox
LEDER CC(O)CNC1[C@H](OCC(C)O)OC(COCC(C)O)[C@@H](O[C@@H]2OC(COCC(C)O)[C@@H](O[C@@H]3OC(COCC(C)O)[C@@H](OCC(C)O)C(OCC(C)O)C3NCC(C)O)C(OCC(C)O)C2NCC(C)O)C1OCC(C)O
Specifikationer
Kemiskt namn eller material Hydroxypropyl Chitosan
Kvantitet 1 g
InChI formel InChI=1S/C51H101N3O24/c1-26(55)12-52-40-47(71-20-34(9)63)44(38(24-67-16-30(5)59)74-49(40)73-22-36(11)65)77-51-42(54-14-28(3)57)48(72-21-35(10)64)45(39(76-51)25-68-17-31(6)60)78-50-41(53-13-27(2)56)46(70-19-33(8)62)43(69-18-32(7)61)37(75-50)23-66-15-29(4)5
Formel vikt 1139.678