Läs mer
Ansell Edmont™ HyFlex™ 11-630 Series Gray Polyurethane Medium Weight Gloves
Handla allt Ansell Produkter
Storlek:
6
7
8
9
10
11
Förpackningsstorlek:
12 par
Beskrivning
- The Lycra™ in the liners of the gloves as well as the polyurethane coating provides excellent flexibility and fit.
- The specific formulation of the polyurethane dipping provides a secure grip on dry or slightly oily pieces.
- The PU formulation also increases abrasion resistance on the gloves’ work surface.
- With strong cut resistance
- Knitted construction
Specifikationer
Specifikationer
| Material på baksidan | Polyurethane |
| Certifikat och efterlevnad | CE Mark, Standard CATEGORY II EN 388 : 4443 |
| Manschettmaterial | Polyurethane |
| Produkttyp | Medium Weight Gloves |
| Storlek | 11 |
| Handske manschettstil | Knitwrist |
| Liner material | Dupont™ Kevlar™ aramid fibers |
| Foder | Nylon/Lycra™/Dyneema™/Cotton/Dupont™ Kevlar™ aramid fiber |
| Typ av palm | Coated-palm |
| Kvantitet | 144 Pairs |
| Visa mer |