Learn More
Ansell™ HyFlex™ 11-925
Click to view available options
Size:
6
7
8
9
10
11
Beschreibung
- The first ultra-light weight HyFlex™ style to combine a ¾ dip geometry, oil repellence and oil grip into a highly dexterous, high comfort glove.
- The HyFlex™ 11-925 features Ansell’s proprietary grip technology providing reduced risk of slippage in dynamic environments.
- The liner is constructed with fine gauge nylon/spandex yarn providing the user with an unrestricted second skin feel.
- This unique blend of protection, performance and flexibility features bridges the gap from previous generations of ¾ dip oil repellent general purpose gloves, delivering enhanced flexibility, tactility, oil grip and extreme comfort.
Spezifikation
Back Material | Nylon, Spandex |
Certifications/Compliance | EN388:2016/3111A |
Classification | Category II |
Coated Area | 3/4 dipped |
Cut Style | Newton |
Description | Ultra-light weight oil repellent, oil grip multi-purpose glove |
Coating Material | Nitrile |
Color | Blue |
Cut Resistance Rating | A |
Disposable | Reusable |
Mehr anzeigen |