Läs mer
Ansell HyFlex™ 11581 Light Duty Touchscreen Compatible Cut-resistant Gloves
Produktkod. 30282354 Handla allt Ansell Produkter
Change view
Klicka för att se tillgängliga alternativ
Storlek:
6
7
8
9
10
11
12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Storlek
30282354 12
30282352 6
30282353 7
30282350 8
30282349 9
30282348 10
30282351 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Produktkod. 30282354 Leverantör Ansell Leverantörsnummer 11581120
Beräknad leverans: 16-10-2026
Lägg i varukorgen
Lägg i varukorgen
missing translation for 'validMainframeMsg'
HyFlex™ 11581 Light Duty Touchscreen Gloves are cut-resistant with a knit wrist cuff, making them ideal for industrial applications.
  • High cut protection with an immediate blade blunting effect outperforming cut protection of gloves in the same category
  • High dexterity and tactility
  • 21-gauge construction makes it the thinnest and lightest glove in its category
  • Touchscreen compatible, so it's ready to use in today's workplaces
  • Extended Use-FORTIX™ technology provides up to 20% more abrasion resistance for longer lasting handling in abrasive conditions
  • Reinforced thumb crotch
  • Made of tungsten, HPPE and nylon
  • Spandex seamless liner
  • DGX nitrile palm coating
TRUSTED_SUSTAINABILITY
Specifikationer
Beläggningsmaterial Nitrile
Färg Gray, Black
Betyg för skärmotstånd ANSI A6
Hand Specific
Latexfri Yes
Material Nylon
Produkttyp Cut-resistant Gloves
Storlek 12