missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Hygiene Kits for Use with Uvex™ K-Series Earmuffs

Click to view available options
Model:
Premium
Standard
For Use With (Equipment):
K1, K1H, and K200 Earmuffs
K2, K2H, K3, and aXess One Earmuffs
This item is not returnable. View return policy
This item is not returnable. View return policy

The Hygiene Kit contains soft memory foam replacement pads for Uvex™ K-Series Earmuffs. It allows timely maintenance of equipment, promoting reuse for good cost management with associated environmental benefits.

  • Product Type: Hygiene Kit
  • For Use With: Uvex™ K-Series Earmuffs
  • Material: Memory Foam
  • Color: Black
  • Models: K1, K1H, K2, K2H, K3, K200, aXess One (specific to kit)
Includes Replacement Pads
For Use With (Equipment) K1, K1H, and K200 Earmuffs
High Visibility No
Model Standard
Product Type Hygiene Kit
Disposable Reusable
Material Memory Foam
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'