Läs mer
Hyoscine Methonitrate, LoGiCal
Produktkod. 30462837
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30462837 10 mg
1 options
Produktkod. 30462837 Leverantör LoGiCal Leverantörsnummer LGCFOR3440.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hyoscine Methonitrate
CAS 6106-46-3
Analyt- eller komponentnamn Hyoscine Methonitrate
Frakt skick Room Temperature
Molekylformel C18 H24 N O4 . N O3
Certifikat och efterlevnad ISO 9001
Synonym Hyoscine Methonitrate, 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 7-[(2S)-3-hydroxy-1-oxo-2-phenylpropoxy]-9,9-dimethyl-, (1alpha,2beta,4beta,5alpha,7beta)-, nitrate (1:1), 1alphaH,5alphaH-Tropanium, 6beta,7beta-epoxy-3alpha-hydroxy-8-methyl-, nitrate, (-)-tropate (8CI), 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 7-(3-hydroxy-1-oxo-2-phenylpropoxy)-9,9-dimethyl-, [7(S)-(1alpha,2beta,4beta,5alpha,7beta)]-, nitrate (salt), 3-Oxa-9-azoniatricyclo[3.3.1.02,4]nonane, 7-[(2S)-3-hydroxy-1-oxo-2-phenylpropoxy]-9,9-dimethyl-, (1alpha,2beta,4beta,5alpha,7beta)-, nitrate (salt) (9CI), N-Methylscopolammonium nitrate (6CI,7CI), 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane, 3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane deriv., Hyoscine methonitrate, Mescomine, Methscopolamine nitrate, Methylscopolamine nitrate, Scopolamine methylnitrate, Skopolate, Skopyl, Viscope
InChI formel InChI=1S/C18H24NO4.NO3/c1-19(2)14-8-12(9-15(19)17-16(14)23-17)22-18(21)13(10-20)11-6-4-3-5-7-11;2-1(3)4/h3-7,12-17,20H,8-10H2,1-2H3;/q+1;-1/t12-,13-,14-,15+,16-,17+;/m1./s1
Lösningstyp Neat
LEDER C[N+]1(C)[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4.[O-][N+](=O)[O-]
Industry Type Clinical Testing
Molekylvikt (g/mol) 380.39
Kvantitet 10 mg
Formel vikt 380.1584 g/mol