Läs mer
Hyoscine N-Oxide Hydrobromide, Mikromol™
Produktkod. 30461990
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461990 4 x 25 mg
30051574 25 mg
30051467 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461990 Leverantör Mikromol Leverantörsnummer MM0546.0700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Hyoscine N-Oxide Hydrobromide, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hyoscine N-Oxide Hydrobromide
CAS 6106-81-6
Analyt- eller komponentnamn Hyoscine N-Oxide Hydrobromide
Frakt skick Room Temperature
Molekylformel C17 H21 N O5 . Br H
Certifikat och efterlevnad ISO 17025
Synonym Hyoscine N-Oxide Hydrobromide
InChI formel InChI=1S/C17H21NO5.BrH/c1-18(21)13-7-11(8-14(18)16-15(13)23-16)22-17(20)12(9-19)10-5-3-2-4-6-10;/h2-6,11-16,19H,7-9H2,1H3;1H/t11-,12-,13-,14+,15-,16+,18?;/m1./s1
Lösningstyp Neat
LEDER Br.C[N+]1([O-])[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)[C@H](CO)c4ccccc4
IUPAC-namn (αS)-α-(Hydroxymethyl)benzeneacetic acid (1α,2β,4β,5α,7β)-9-methyl-3-oxa-9-oxido-9-azatricyclo[3.3.1.02,4]non-7-yl ester hydrobromide
Molekylvikt (g/mol) 400.26
Kvantitet 4 x 25 mg
Formel vikt 399.0681 g/mol