Läs mer
Safety First Aid HypaClens Emergency Eye Wash Station, Value (2 x 500 mL Bottles)
Beskrivning
- It is ideal for short-term construction projects, especially where no clean running water is available.
- HypaClens Sterile Eyewash is a single-use medical device intended for emergency eye irrigation.
- As per HSE guidance, the eye should be washed out with either clean water or sterile fluid from a sealed container for at least 10 min. following contamination from chemicals or small particles.
- The eyewash is designed as a substitute for clean running water in first aid situations and is only intended to be used once.
- Re-use of the bottle would mean you can no longer guarantee the contents are sterile and face the risk of infection.
- For this reason, the bottle is designed so it cannot be resealed.
- After use, any excess fluid remaining should be discarded.
Specifikationer
Specifikationer
| Färg | Clear |
| Flödeshastighet | 20 gpm (Shower), 8.0 gpm (Eye and Face Wash) |
| Material | Plastic |
| Montering | Wall |
| Produkttyp | Portable Eyewash and Shower Fixtures |