Learn More
Thermo Scientific™ HyperSep™ Pressure Manifold Air Filter
Prevent contaminants in the air source from entering the manifold system.
Brand: Thermo Scientific™ 60104-273
Additional Details : Weight : 1.00000kg
Description
Prevent contaminants from the air source from entering the system the Thermo Scientific™ HyperSep™ Positive Pressure Manifold Installation Kit for all the parts needed to set up your manifold. The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes.
See related products if you need replacements for any of these HperSep Positive Pressure Manifold accessories.Specifications
In Line Air Filter, 1 pk | |
Positive Pressure Manifold | |
In Line Air Filter |
SPE | |
HyperSep |