Läs mer
Thermo Scientific™ HyperSep™ Servo Cocaine SPE Cartridges and Well Plates
Produktkod. 11312443 Handla allt Thermo Scientific Produkter
Beskrivning:
1 g/6 ml SPE-kolonn 30-pack
130 mg/1 ml SPE-kolonn 100-pack
200 mg/6 ml SPE-kolonn 50-pack
300 mg/3 ml SPE-kolonn 50-pack
500 mg/3 ml SPE-kolonn 50-pack
96-brunnsplatta 10 mg 1 förpackning
96-brunnsplatta 100 mg 1 förpackning
96-brunnsplatta 25 mg 1 förpackning
96-brunnsplatta 50 mg 1 förpackning
Förpackningsstorlek:
Paket med 100
Paket med 30
Paket med 50
Varje
Produktkod. 11312443
Brand: Thermo Scientific™ 60111701C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
For high confidence screening for drugs of abuse, these SPE cartridges and plates offer a robust, ready-to-use solution for extraction of cocaine class compounds.
Get the cleanest possible sample extracts—and maximum confidence—when preparing samples for screening for drugs of abuse. Depending on your analytical needs, you can choose from a comprehensive range of Thermo Scientific™ HyperSep Servo products, each optimized for a different class of drugs. To ensure success, each product comes with detailed extraction procedures.
Thermo Scientific™ HyperSep Servo Cocaine Cartridges and Well Plates offer a robust, ready-to-use solution for dedicated extraction of cocaine class compounds.
HyperSep Servo and Servo+ Cartridges and Plates
Thermo Scientific™ HyperSep Servo and Servo+ cartridges and plates provide complete, high-reliability SPE sample preparation solutions for drug screening applications.
Both Servo and Servo+ product lines deliver the high levels of reproducibility and clean sample extracts required for maximum confidence when dealing with complex biological sample matrices. Servo+ cartridges and plates offer an extra measure of selectivity, higher loading capacities and enhanced robustness.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifikationer
Beskrivning 130 mg/1 ml SPE-kolonn 100-pack
Sängvikt 130 mg
För användning med (applikation) Analysis of Drugs of Abuse
Produktlinje HyperSep
Sorbent HyperSep Servo Cocaine
Typ Solid Phase Extraction Column
Volym (metrisk) 1 mL