Läs mer
Hypromellose Acetate Succinate, TRC
Produktkod. 30403872
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30403872 5 g
1 options
Produktkod. 30403872 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9987935G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hypromellose Acetate Succinate
CAS 71138-97-1
Molekylformel C10H22O9
Kvantitet 5 g
Synonym Hypromellose ASLF, Affinisol HPMCAS 912G
InChI formel InChI=1S/C4H6O4.C3H8O2.C2H4O2.CH4O/c5-3(6)1-2-4(7)8;1-3(5)2-4;1-2(3)4;1-2/h1-2H2,(H,5,6)(H,7,8);3-5H,2H2,1H3;1H3,(H,3,4);2H,1H3
LEDER CC(CO)O.CC(=O)O.CO.C(CC(=O)O)C(=O)O
Molekylvikt (g/mol) 286.28