Läs mer
Tocris Bioscience™ I-BRD9
Potent and selective BRD9 inhibitor
Brand: Tocris Bioscience™ 5591/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
I-BRD9 is a potent and selective BRD9 inhibitor (pIC50 = 7.3). Exhibits >70-fold selectivity for BRD9 over a panel of 34 other bromodomains and >700-fold selectivity over the BET family. Downregulates CLEC1, DUSP6, FES and SAMSN1 genes in Kasumi-1 cells.Specifikationer
| 5-Ethyl-4,5-dihydro-4-oxo-N-(tetrahydro-1,1-dioxido-2H-thiopyran-4-yl)-7-[3-(trifluoromethyl)phenyl]thieno[3,2-c]pyridine-2-carboximidamide | |
| 1714146-59-4 | |
| 10 mg |