Learn More
Invitrogen™ Silencer™ Human Kinase siRNA Library
Product Code. 15681842
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
Product Code. Quantity Unit Size
15681842 1 Set 1 set
1 options
Product Code. 15681842 Supplier Invitrogen™ Supplier No. A30079
 Restricted Item
Beräknad leverans: 07-12-2026
Given their importance in cell signaling and many other biological pathways, kinases have become key drug targets.
Given their importance in cell signaling and many other biological pathways, kinases have become key drug targets. The Silencer™ Human Kinase siRNA Library contains 2127 Ambion™ Silencer siRNAs targeting 709 human kinase genes for performing RNAi screens against the kinome. This siRNA library is designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC) as well as additional resources, such as the KinBase database. Utilizing Ambion Silencer technology built on stringent bioinformatics to enable high target specificity and siRNA potency, the Silencer Human Kinase siRNA Library allows accurate yet cost-effective RNAi screening for kinase signaling pathways.
  • Comprehensive – Expedite functional gene discovery with pre-defined Silencer Human Kinase siRNA Library
  • Reliable – Increase target screening accuracy with three individual siRNAs per gene target
  • Efficient – Reduce off target impacts at low concentrations with proven Ambion Silencer siRNA design
  • Convenient – Reduce data de-convolution and accelerate target validation with our arrayed format
  • Customizable – Choose the best option available for your workflow demands, including additional siRNA scales and pooled formats
Why use the Silencer Human Kinase siRNA Library?
Silencer siRNAs are optimized for design and high quality synthesis with guaranteed silencing. The Silencer Human Kinase siRNA Library contains our pre-designed and when available, validated siRNAs generated with potency and specificity in mind so you can focus on data quality. This library is selected against the latest available databases and resources for up-to-date content. Full siRNA sequence information, as well as the exon targeted and each targeted gene's symbol, full name, aliases, RefSeq number, and Locus ID, plus results from any validation experiments performed using the included siRNAs is provided. Please contact us for the complete list of targets.
Silencer siRNA libraries allow you to screen efficiently and maximize data output
Ambion Silencer siRNAs are designed with potency in mind to help ensure gene knockdown at minimum concentrations. This not only helps ensure less potential for off target effects and false positive data but also minimizes false negative outcomes of siRNA screening. The Silencer Human Kinase siRNA Library contains 0.25 nmol of each siRNA, sufficient for 100 transfections (at 25 nM siRNA, 100 μL transfection volume).
Flexibility to meet your workflow needs
Thermo Fisher Scientific offers comprehensive list of pre-defined or custom siRNA libraries to human, mouse or rat genes with 1, 2, or 5 nmol of each siRNA or larger sizes. For a complete gene list, additional information or to discuss your research with one of our technical experts, contact your local sales representative or e-mail us at RNAilibraries@lifetech.com.
TRUSTED_SUSTAINABILITY
Specifications
Description 2127 total siRNAs targeting 709 genes plated onto 96-well plates (27 plates)
Content And Storage 2127 total siRNAs targeting 709 genes plated onto 27 x 96-well plates
Store at or below -20°C in a non-frost-free freezer.
Format 96-well Plate
RNAi Type siRNA
Species Human
For Use With (Application) RNAi
Product Line Silencer
Product Type siRNA Library
Purity Standard
Quantity 1 Set
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice