Beskrivning
- The lightweight, sporty design and panoramic lens make the eyewear extremely comfortable to wear - even over long periods of time.
- The outstanding, long-lasting uvex supra-vision excellence coating technology, uvex i-works safety spectacles boast long-lasting anti-fogging properties that withstand repeated cleaning.
- Comfortable, soft duo-component side arms ensure an easy-to-wear, non-slip fit.
- Sporty, rugged safety spectacles
- Available with different lens tints
- Robust polycarbonate lens with proven uvex supra-vision coating technology - permanently anti-fog and scratch-resistant
- Fits the uvex IES Better for the environment, since lenses can be replaced as necessary
- Metal-free
- Ergonomically shaped side arms with soft ear-pieces ensure a comfortable, secure fit without creating pressure points
- Anatomically shaped nose piece for all-day wearer comfort
- Certified in accordance with EN 166 (personal eye protection) and EN 170 (UV filters) UV400 - 100% protection against harmful UV radiation up to 400 nm
Specifikationer
Specifikationer
| Linsbeläggning | Anti-fog, Anti-scratch, Scratch-resistant |
| Linsfärg | Clear |
| Produkttyp | Safety Glasses |
| Material | Polycarbonate |
| Disponibel | Reusable |
| Linsmaterial | Polycarbonate |
| Rammaterial | TPU |
| UV-skydd | UV400 |
| Ramtyp | Semi-rimless |