Learn More
IA-6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560857
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KHLSTHEAGS ARALAPGFGS ERGAPLAFAC PLCGAHFPTA DIREKHRLWH AVREELLLPA LAGAPPETSG PSGPSDGSAQ QIFSCKHCPS TF Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.
IA-6 Polyclonal antibody specifically detects IA-6 in Human samples. It is validated for Immunohistochemistry.Specifications
IA-6 | |
Polyclonal | |
Unconjugated | |
INSM2 | |
IA6; IA-6; INSM transcriptional repressor 2; Insm2; INSM2 variant 1; insulinoma-associated 2; insulinoma-associated protein 2; insulinoma-associated protein IA-6; methylated in liver tumor 1; mLt 1; mLt1; Nbla106; Zinc finger protein IA-6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84684 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96T92 | |
INSM2 | |
Recombinant protein corresponding to Human IA-6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only