Läs mer
IACS-010759, MedChemExpress
Produktkod. 16485396
Encompass_Preferred
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
16485396 100 mg
1 options
Produktkod. 16485396 Leverantör Medchem Express Leverantörsnummer HY112037
MedChemExpress IACS-010759 is an orally active, potent mitochondrial complex I of oxidative phosphorylation (OXPHOS) inhibitor. IACS-010759 inhibits proliferation and induces apoptosis in models of brain cancer and acute myeloid leukemia (AML) reliant on OXPHOS. IACS-010759 has the potential for relapsed/refractory AML and solid tumors research.
IACS-010759 is an orally active, potent mitochondrial complex I of oxidative phosphorylation (OXPHOS) inhibitor. IACS-010759 inhibits proliferation and induces apoptosis in models of brain cancer and acute myeloid leukemia (AML) reliant on OXPHOS. IACS-010759 has the potential for relapsed/refractory AML and solid tumors research.
Specifikationer
Kemiskt namn eller material IACS-010759
CAS 1570496-34-2
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C25H25F3N6O4S
Kvantitet 100 mg
Synonym IACS-10759
Löslighetsinformation DMSO : 62.5 mg/mL (111.10 mM; Need ultrasonic)
LEDER FC(F)(F)OC1=CC=C(C2=NOC(C3=NN(CC4=CC(N5CCC(S(=O)(C)=O)CC5)=CC=C4)C(C)=N3)=N2)C=C1
Molekylvikt (g/mol) 562.56
Formel vikt 562.56
Procent renhet 98.7%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease