missing translation for 'onlineSavingsMsg'
Läs mer

IACS-8968, MedChemExpress

Produktkod. 16497297
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16497297 5 mg
16444588 10 mg
16431169 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16497297 Leverantör Medchem Express Leverantörsnummer HY112164

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress IACS-8968 (IDO/TDO Inhibitor) is a dual IDO and TDO inhibitor, with pIC50s of 6.43 for IDO and <5 for TDO, respectively.

IACS-8968 (IDO/TDO Inhibitor) is a dual IDO and TDO inhibitor, with pIC50s of 6.43 for IDO and <5 for TDO, respectively.

Specifikationer

Kemiskt namn eller material IACS-8968
CAS 2144425-14-7
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C17H18F3N5O2
Kvantitet 5 mg
Synonym IDO/TDO Inhibitor
Löslighetsinformation DMSO : 50 mg/mL (131.11 mM; Need ultrasonic)
LEDER O=C(N1)NC2(C(C)(C)CN(C3=CC(C(F)(F)F)=CC4=CN=CN34)CC2)C1=O
Molekylvikt (g/mol) 381.35
Formel vikt 381.35
Procent renhet 98.29%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death
Visa mer Visa mindre

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.