missing translation for 'onlineSavingsMsg'
Läs mer

(±)-Ibipinabant, MedChemExpress

Produktkod. 16486168
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
50 mg
Förpackningsstorlek:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16486168 10 mg 10mg
16468767 5 mg -
16462307 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16486168 Leverantör Medchem Express Leverantörsnummer HY14791A

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress (±)-Ibipinabant ((±)-SLV319) is the racemate of SLV319. (±)-Ibipinabant ((±)-SLV319) is a potent and selective cannabinoid-1 (CB-1) receptor antagonist with an IC50 of 22 nM.

(±)-Ibipinabant ((±)-SLV319) is the racemate of SLV319. (±)-Ibipinabant ((±)-SLV319) is a potent and selective cannabinoid-1 (CB-1) receptor antagonist with an IC50 of 22 nM.

Specifikationer

Kemiskt namn eller material (±)-Ibipinabant
CAS 362519-49-1
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C23H20Cl2N4O2S
Kvantitet 10 mg
Synonym (±)-SLV319 (±)-BMS-646256
Löslighetsinformation DMSO : ≥ 31 mg/mL (63.60 mM)
LEDER C/N=C(N1N=C(C2=CC=C(Cl)C=C2)C(C3=CC=CC=C3)C1)\NS(=O)(C4=CC=C(Cl)C=C4)=O
Molekylvikt (g/mol) 487.4
Formel vikt 487.4
Procent renhet 99.93%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Metabolism-sugar/lipid metabolism
Visa mer Visa mindre

Research purposes only

Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.