Läs mer
Ibrutinib Dimer (~90%), TRC
Produktkod. 30136816
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136816 10 mg
30135913 25 mg
30008328 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136816 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI12498510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2031255-23-7
Molekylformel C50H48N12O4
Molekylvikt (g/mol) 880.99
Synonym 1-((R)-3-(4-((3-((R)-3-(4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidin-1-yl)-3-oxopropyl)amino)-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidin-1-yl)prop-2-en-1-one
IUPAC-namn 1-[(3R)-3-[4-amino-3-(4-phenoxyphenyl)pyrazolo[3,4-d]pyrimidin-1-yl]piperidin-1-yl]-3-[[3-(4-phenoxyphenyl)-1-[(3R)-1-prop-2-enoylpiperidin-3-yl]pyrazolo[3,4-d]pyrimidin-4-yl]amino]propan-1-one
LEDER C=CC(N1CCC[C@@H](N2C3=NC=NC(NCCC(N4CCC[C@@H](N5C6=NC=NC(N)=C6C(C7=CC=C(OC8=CC=CC=C8)C=C7)=N5)C4)=O)=C3C(C9=CC=C(OC%10=CC=CC=C%10)C=C9)=N2)C1)=O
Specifikationer
Kemiskt namn eller material Ibrutinib Dimer
Kvantitet 10 mg
InChI formel InChI=1S/C50H48N12O4/c1-2-41(63)59-27-9-11-35(29-59)62-50-44(46(58-62)34-19-23-40(24-20-34)66-38-15-7-4-8-16-38)48(54-32-56-50)52-26-25-42(64)60-28-10-12-36(30-60)61-49-43(47(51)53-31-55-49)45(57-61)33-17-21-39(22-18-33)65-37-13-5-3-6-14-37/h2-8,13-24,31-32,35-36H,1,9-12,25-30H2,(H2,51,53,55)(H,52,54,56)/t35-,36-/m1/s1
Formel vikt 880.3921