Läs mer
Ibufenac, LoGiCal
Produktkod. 30464078
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30464078 10 mg
1 options
Produktkod. 30464078 Leverantör LoGiCal Leverantörsnummer LGCFOR1872.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ibufenac
CAS 1553-60-2
Analyt- eller komponentnamn Ibufenac
Frakt skick Room Temperature
Molekylformel C12 H16 O2
Certifikat och efterlevnad ISO 9001
Synonym Benzeneacetic acid, 4-(2-methylpropyl)-, Acetic acid, (p-isobutylphenyl)- (8CI), (p-Isobutylphenyl)acetic acid, 4-(2-Methylpropyl)benzeneacetic acid, 4-(2-Methylpropyl)phenylacetic acid, 4-Isobutoxyphenylacetic acid, 4-Isobutylphenylacetic acid, Dytransin, Ibufenac, Ibunac, Isodilan, Medirex, NSC 99976, RD 11654
InChI formel InChI=1S/C12H16O2/c1-9(2)7-10-3-5-11(6-4-10)8-12(13)14/h3-6,9H,7-8H2,1-2H3,(H,13,14)
Lösningstyp Neat
LEDER CC(C)Cc1ccc(CC(=O)O)cc1
IUPAC-namn 2-[4-(2-methylpropyl)phenyl]acetic acid
Industry Type Clinical Testing
Molekylvikt (g/mol) 192.25
Kvantitet 10 mg
Formel vikt 192.115 g/mol