Läs mer
Ibufenac, TRC
Produktkod. 30179833
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179833 100 mg
30180031 50 mg
30179993 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179833 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI130000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1553-60-2
Molekylformel C12 H16 O2
Molekylvikt (g/mol) 192.25
Synonym Benzeneacetic acid, 4-(2-methylpropyl)-, Acetic acid, (p-isobutylphenyl)- (8CI), (p-Isobutylphenyl)acetic acid, 4-(2-Methylpropyl)benzeneacetic acid, 4-(2-Methylpropyl)phenylacetic acid, 4-Isobutoxyphenylacetic acid, 4-Isobutylphenylacetic acid, Dytransin, Ibufenac, Ibunac, Isodilan, Medirex, NSC 99976, RD 11654
IUPAC-namn 2-[4-(2-methylpropyl)phenyl]acetic acid
LEDER CC(C)Cc1ccc(CC(=O)O)cc1
Specifikationer
Kemiskt namn eller material Ibufenac
Kvantitet 100 mg
InChI formel InChI=1S/C12H16O2/c1-9(2)7-10-3-5-11(6-4-10)8-12(13)14/h3-6,9H,7-8H2,1-2H3,(H,13,14)
Formel vikt 192.115