Läs mer
Ibuprofen Related Compound (2,3-Dihydroxypropyl 2-(4-Isobutylphenyl)Propanoate), TRC
Produktkod. 30137172
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137172 50 mg
30132619 100 mg
17951695 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137172 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI40014550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 64622-21-5
Molekylformel C16 H24 O4
Molekylvikt (g/mol) 280.36
Synonym 2,3-Dihydroxypropyl 2-(4-Isobutylphenyl)propanoate
IUPAC-namn 2,3-dihydroxypropyl 2-[4-(2-methylpropyl)phenyl]propanoate
LEDER CC(C)Cc1ccc(cc1)C(C)C(=O)OCC(O)CO
Specifikationer
Kemiskt namn eller material 2,3-Dihydroxypropyl 2-(4-Isobutylphenyl)propanoate
Kvantitet 50 mg
InChI formel InChI=1S/C16H24O4/c1-11(2)8-13-4-6-14(7-5-13)12(3)16(19)20-10-15(18)9-17/h4-7,11-12,15,17-18H,8-10H2,1-3H3
Formel vikt 280.1675