Läs mer
IC261, MedChemExpress
Produktkod. 16465338
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16465338 10 mg
16438057 5 mg
16431537 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16465338 Leverantör Medchem Express Leverantörsnummer HY12774
MedChemExpress IC261 is a selective, ATP-competitive CK1 inhibitor, with IC50s of 1 μM, 1 μM, 16 μM for Ckiδ, Ckiε and Ckiα1, respectively.
IC261 is a selective, ATP-competitive CK1 inhibitor, with IC50s of 1 μM, 1 μM, 16 μM for Ckiδ, Ckiε and Ckiα1, respectively.
Specifikationer
Kemiskt namn eller material IC261
CAS 186611-52-9
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C18H17NO4
Kvantitet 10 mg
Löslighetsinformation DMSO : ≥ 33 mg/mL (106.00 mM)
LEDER O=C1NC2=C(C=CC=C2)/C1=C\C3=C(OC)C=C(OC)C=C3OC
Molekylvikt (g/mol) 311.33
Formel vikt 311.33
Procent renhet 99.75%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease