Läs mer
Tocris Bioscience™ ICA 110381
GREENER_CHOICE
Produktkod. 30001604
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30001604 50 mg
30001582 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30001604 Leverantör Tocris Bioscience™ Leverantörsnummer 4950/50
KV7.2/7.3 activator; displays anticonvulsant properties
KV7.2/7.3 activator (EC50 = 0.38 μM). Decreases neuronal excitability in CA1 hippocampal neurons. Exhibits anticonvulsive properties in amygdala-kindled rats, a model for complex partial seizures.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 325457-99-6
Molekylformel C12H8Cl2N2O
Specifikationer
Kemiskt namn eller material ICA 110381
Kvantitet 50 mg
Formel vikt 267.11
Renhet >98%
Molekylvikt (g/mol) 267.11