missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICA1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18021-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
ICA1L | |
Polyclonal | |
Unconjugated | |
ICA1L | |
ALS2CR14, ALS2CR15, ICA1L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
130026, 316432, 70375 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3TY65, Q6RUG5, Q8NDH6 | |
Ica1l | |
ICA1L Fusion Protein Ag12570 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |