missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Icam-1 Rabbit anti-Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16174-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specifically against mouse Icam-1.
Intercellular adhesion molecule-1 (ICAM-1, CD54) is an integral membrane protein, a member of the immunoglobulin superfamily, and a ligand for lymphocyte function-associated (LFA) antigens, a beta 2 leukocyte integrin. The normal function of human ICAM-1 is to provide adhesion between endothelial cells and leukocytes after injury or stress. ICAM-1 binds to leukocyte function-associated antigen (LFA-1) or macrophage-1 antigen (Mac-1). It is found on leukocytes, fibroblasts, epithelial cells and endothelial cells and its expression is regulated by inflammatory cytokines. ICAM-1 has a tissue distribution similar to that of the major histocompatibility complex class II antigens and is likely to play a role in inflammatory responses.Specifications
Icam-1 | |
Polyclonal | |
Unconjugated | |
Icam1 | |
CD54, ICAM 1, Icam1, Ly 47, MALA 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15894, 25464 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P13597, Q00238 | |
Icam1 | |
Peptide | |
150 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |