Learn More
ICAM2 Mouse anti-Human, Unconjugated, Clone: 1H3C6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67628-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The ICAM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ICAM2. This antibody recognizes human antigen. The ICAM2 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
ICAM2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Flow Cytometry, ELISA | |
1H3C6 | |
Unconjugated | |
20 μL | |
3384 | |
IgG2a |