Learn More
ICAM2 Mouse anti-Human, Unconjugated, Clone: 1H3C6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67628-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16800366
  • 1656.73 SEK / 20µL
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
The ICAM2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ICAM2. This antibody recognizes human antigen. The ICAM2 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
Specifications
ICAM2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Flow Cytometry, ELISA
1H3C6
Unconjugated
20 μL
3384
IgG2a