missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ICAM3 (Human) Recombinant Protein
Human ICAM3 full-length ORF ( AAH58903, 30 a.a. - 547 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003385-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: QEFLLRVEPQNPVLSAGGSLFVNCSTDCPSSEKIALETSLSKELVASGMGWAAFNLSNVTGNSRILCSVYCNGSQITGSSNITVYRLPERVELAPLPPWQPVGQNFTLRCQVEDGSPRTSLTVVLLRWEEELSRQPAVEEPAEVTATVLASRDDHGAPFSCRTELDMQPQGLGLFVNTSAPRQLRTFVLPVTPPRLVAPRFLEVETSWPVDCTLDGLFPASEAQVYLALGDQMLNATVMNHGDTLTATATATARADQEGAREIVCNVTLGGERREARENLTVFSFLGPIVNLSEPTAHEGSTVTVSCMAGARVQVTLDGVPAAAPGQPAQLQLNATESDDGRSFFCSATLEVDGEFLHRNSSVQLRVLYGPKIDRATCPQHLKWKDKTRHVLQCQARGNPYPELRCLKEGSSREVPVGIPFFVNVTHNGTYQCQASSSRGKYTLVVVMDIEAGSSHFVPVFVAVLLTLGVVTIVLALMYVFREHQRSGSYHVREESTYLPLTSMQPTEAMGEEPSRAE
Specifications
AAH58903 | |
intercellular adhesion molecule 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ICAM3 | |
GST |
3385 | |
Wheat germ expression system | |
25 μg | |
CD50, CDW50, ICAM-R | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |