Läs mer
Icatibant Acetate, TRC
Produktkod. 17901295
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17901295 25 mg
17902815 5 mg
17964465 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17901295 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI16375025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 138614-30-9
Synonym L-Arginine, D-arginyl-L-arginyl-L-prolyl-(4R)-4-hydroxy-L-prolylglycyl-3-(2-thienyl)-L-alanyl-L-seryl-(3R)-1,2,3,4-tetrahydro-3-isoquinolinecarbonyl-(2S,3aS,7aS)-octahydro-1H-indole-2-carbonyl-, acetate (salt) (9CI)∣L-Arginine, D-arginyl-L-arginyl-L-prolyl-trans-4-hydroxy-L-prolylglycyl-3-(2-thienyl)-L-alanyl-L-seryl-D-1,2,3,4-tetrahydro-3-isoquinolinecarbonyl-L-(2α,3aβ,7aβ)-octahydro-1H-indole-2-carbonyl-, acetate (salt)∣Firazyr∣HOE 140∣D-Arginyl-L-arginyl-L-prolyl-(4R)-4-hydroxy-L-prolylglycyl-3-(2-thienyl)-L-alanyl-L-seryl-(3R)-1,2,3,4-tetrahydro-3-isoquinolinecarbonyl-(2S,3aS,7aS)-octahydro-1H-indole-2-carbonyl-L-arginine Acetate Salt∣D-Arginyl-L-arginyl-L-prolyl-trans-4-hydroxy-L-prolylglycyl-3-(2-thienyl)-L-alanyl-L-seryl-D-1,2,3,4-tetrahydro-3-isoquinolinecarbonyl-L-(2α,3aβ,7aβ)-octahydro-1H-indole-2-carbonyl-L-arginine Acetate Salt
Specifikationer
Formatera Neat
Molekylvikt (g/mol) 1364.6
Produkttyp API