Läs mer
Icatibant Acetate, TRC
Produktkod. 17964465
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17964465 1 mg
17901295 25 mg
17902815 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17964465 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI1637501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 138614-30-9
Synonym L-Arginine, D-arginyl-L-arginyl-L-prolyl-(4R)-4-hydroxy-L-prolylglycyl-3-(2-thienyl)-L-alanyl-L-seryl-(3R)-1,2,3,4-tetrahydro-3-isoquinolinecarbonyl-(2S,3aS,7aS)-octahydro-1H-indole-2-carbonyl-, acetate (salt) (9CI)∣L-Arginine, D-arginyl-L-arginyl-L-prolyl-trans-4-hydroxy-L-prolylglycyl-3-(2-thienyl)-L-alanyl-L-seryl-D-1,2,3,4-tetrahydro-3-isoquinolinecarbonyl-L-(2α,3aβ,7aβ)-octahydro-1H-indole-2-carbonyl-, acetate (salt)∣Firazyr∣HOE 140∣D-Arginyl-L-arginyl-L-prolyl-(4R)-4-hydroxy-L-prolylglycyl-3-(2-thienyl)-L-alanyl-L-seryl-(3R)-1,2,3,4-tetrahydro-3-isoquinolinecarbonyl-(2S,3aS,7aS)-octahydro-1H-indole-2-carbonyl-L-arginine Acetate Salt∣D-Arginyl-L-arginyl-L-prolyl-trans-4-hydroxy-L-prolylglycyl-3-(2-thienyl)-L-alanyl-L-seryl-D-1,2,3,4-tetrahydro-3-isoquinolinecarbonyl-L-(2α,3aβ,7aβ)-octahydro-1H-indole-2-carbonyl-L-arginine Acetate Salt
Specifikationer
Formatera Neat
Molekylformel C61H93N19O15S
Molekylvikt (g/mol) 1364.6, 1364.59
InChI-nyckel HKMZRZUEADSZDQ-QNQUGUDESA-N
LEDER CC(O)=O.N[C@H](CCCN=C(N)N)C(=O)NC(CCCN=C(N)N)C(=O)N1CCCC1C(=O)N1C[C@H](O)CC1C(=O)NCC(=O)N[C@@H](CC1=CC=CS1)C(=O)N[C@@H](CO)C(=O)N1CC2=CC=CC=C2CC1C(=O)N1[C@H]2CCCC[C@H]2CC1C(=O)N[C@@H](CCCN=C(N)N)C(O)=O
IUPAC-namn (2S)-2-{[(3aS,7aS)-1-{2-[(2S)-2-[(2S)-2-(2-{[(4R)-1-(1-{2-[(2R)-2-amino-5-[(diaminomethylidene)amino]pentanamido]-5-[(diaminomethylidene)amino]pentanoyl}pyrrolidine-2-carbonyl)-4-hydroxypyrrolidin-2-yl]formamido}acetamido)-3-(thiophen-2-yl)propanamido]-3-hydroxypropanoyl]-1,2,3,4-tetrahydroisoquinoline-3-carbonyl}-octahydro-1H-indol-2-yl]formamido}-5-[(diaminomethylidene)amino]pentanoic acid; acetic acid
Produkttyp API